accessibility menu, dialog, popup

UserName

(R)S(S)(S)(S)(R)(S)(S)(R)(R)OOOO(R)NHOHOH(R)HOHHHHNaMeMeMe

CAS RN 345909-26-4

IUPAC Name: sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]ethane-1-sulfonate
Synonyms: sodium taurocholate|taurocholate sodium|taurocholic acid sodium salt|taurocholate sodium salt|monosodium N-choloyltaurinate
Molecular Weight (g/mol): 537.69
Molecular Formula: C26H44NNaO7S
InChi Key: JAJWGJBVLPIOOH-IZYKLYLVSA-M
SMILES: [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC(=O)NCCS([O-])(=O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results