missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tartrate, 10% (w/v), Ricca Chemical

Catalog No. 785016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
785016 500 mL Natural Poly Bottle
785032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 785016 Supplier Ricca Chemical Company Supplier No. 785016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6106-24-7 , 7732-18-5
Molecular Formula C4H10Na2O8
Molecular Weight (g/mol) 232.096
InChI Key YAPIZISBZUXUIH-DGFHWNFOSA-N
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
SMILES C(C(C(=O)O)O)(C(=O)O)O.O.O.[Na].[Na]

Specifications

CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Sodium Tartrate