missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, Reagent, ACS, 98%, Spectrum™ Chemical

Catalog No. 18606864
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606864 50 kg Poly Bottle
18606861 2.5 kg Poly Bottle
18606860 6 x 500 g Poly Bottle
18606862 4 x 2.5 kg Poly Bottle
18606863 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606864 Supplier Spectrum Chemical Mfg Cor Supplier No. S147550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1475, 7757-83-7, Na2SO3

Spectrum™ Chemical Sodium Sulfite, Reagent, ACS is a used for preserving meat and prevent discoloration of dried fruit. It is a soluble sodium salt of sulfuric acid. It is also used in the paper processing industry and to purify TNT. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula H14Na2O10S
Molecular Weight (g/mol) 252.14
InChI Key WMJRPJZQQSSDBU-UHFFFAOYSA-L
IUPAC Name disodium heptahydrate sulfite
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 50 kg
Percent Purity 98%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Sulfite