missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, puriss., Honeywell Fluka™

Catalog No. 6001998
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001998 2.5 kg
60044813 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001998 Supplier Honeywell Chemicals Supplier No. 134712.5KG
Only null left

Puriss., Meets analytical specification of BP, Ph. Eur., NF, Anhydrous, 95.0-100.0% (iodometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Anhydrous
Quantity 2.5 kg
Linear Formula Na2SO3
UN Number NONH for all modes of transport
Formula Weight 126.04g/mol
Percent Purity 95 to 100% (iodometric)
Grade puriss.
Chemical Name or Material Sodium Sulfite, Meets analytical specification of Ph.Eur.BP, NF
EINECSNumber 231-821-4
RTECSNumber WE2150000