missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, Exsiccated, Purified, Spectrum™ Chemical

$303.25 - $3373.94

Chemical Identifiers

CAS 7757-83-7
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613886
View Documents
Spectrum Chemical Mfg Cor
S1477500GM
500 g Poly Bottle
Each for $303.25
Only null left
 
18613887
View Documents
Spectrum Chemical Mfg Cor
S147725KG
2.5 kg Poly Pail
Each for $778.59
Only null left
 
18613888
View Documents
Spectrum Chemical Mfg Cor
S147712KG
12 kg Poly Pail
Each for $1,831.72
Only null left
 
18613889
View Documents
Spectrum Chemical Mfg Cor
S147745KGBL
45 kg Fiber Drum
Each for $3,373.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7757-83-7

Specifications

7757-83-7
500 g
Poly Bottle
1
Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.