Learn More
Sodium Sulfite Anhydrous (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific S4303
Specifications
| Sodium Sulfite Anhydrous | |
| 7757-83-7 | |
| 8.5 to 10 | |
| 3 kg | |
| Na2O3S | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| [O-]S(=O)[O-].[Na+].[Na+] | |
| 126.037 | |
| CHEBI:86477 | |
| ≥98% | |
| Pass Test | |
| >500°C |
| 500°C | |
| White | |
| Solid | |
| ≥98 % | |
| MFCD00003503 | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| disodium;sulfite | |
| 24437 | |
| 126.04 | |
| Certified ACS | |
| Glass Bottle | |
| 2.630g/cm³ |
Chemical Identifiers
| 7757-83-7 | |
| 126.037 | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| 24437 | |
| disodium;sulfite |
| Na2O3S | |
| MFCD00003503 | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| CHEBI:86477 | |
| [O-]S(=O)[O-].[Na+].[Na+] |
Safety and Handling
WARNING!
Emergency Overview
Contact with acids liberates toxic gas. May cause central nervous system effects. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:0
Instability:1