missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, Anhydrous, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCS0870500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0870500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS0870500 Supplier Ricca Chemical Company Supplier No. RDCS0870500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals or Granules
Quantity 500 g
Percent Purity ≥ 98%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Sulfite, Anhydrous
ShelfLife 30 months