missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite Anhydrous ACS, MilliporeSigma™

Catalog No. p-3176506
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX07851 500 g Poly Bottle
MSX07853 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MSX07851 Supplier Technidata Supplier No. SX07851
Only null left

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Quantity 500 g
Linear Formula Na2SO3
Formula Weight 126.04
Grade ACS
Packaging Poly Bottle
Chemical Name or Material Sodium Sulfite, Anhydrous