missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, Anhydrous, 98.0% min., ACS, MilliporeSigma™

Catalog No. MSX07855
Change view
Click to view available options
Quantity:
12 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX07855 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX07855 Supplier Technidata Supplier No. SX07855

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity ACS reagent grade sodium sulfite suitable for laboratory analysis and testing applications.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Powder
Quantity 12 kg
Assay Percent Range 98.0% min.
Formula Weight 126.04
Percent Purity ≥98%
Grade ACS
Chemical Name or Material Sodium Sulfite, Anhydrous, For ACS analysis