missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfite, ACS Grade, ≥ 98.0%, LabChem™
Specifications
Specifications
| Melting Point | 500°C |
| Color | White |
| CAS Max % | 100 |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Linear Formula | Na2SO3 |
| Solubility Information | Soluble in water |
| Formula Weight | 126.04 |
| Percent Purity | ≥98.0% |
| Grade | ACS |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| EINECSNumber | 231-821-4 |
| Recommended Storage | Room Temperature |