missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite ACS AR Granular, Macron Fine Chemicals™

Catalog No. MK8064500
Change view
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly
POLYSTORMOR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK8064500 500g POLYSTORMOR
MK8064212 2.5kg Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK8064500 Supplier Avantor Supplier No. 806404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Quantity 500g
Formula Weight 126.04
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Sodium Sulfite, Anhydrous