missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, ACS, 98.0% min

Catalog No. AA13454A9
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13454A9 10 kg
AA1345436 500 g
AA13454A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA13454A9 Supplier Thermo Scientific Chemicals Supplier No. 013454A9

Sodium sulfite is used in the photography industry to protect developing solutions from oxidation; in the pulp and paper industry; in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in the textile industry. | CAS: 7757-83-7 | Na2O3S | 126.037 g/mol

Sodium sulfite is used in the photography industry to protect developing solutions from oxidation; in the pulp and paper industry; in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in the textile industry. It is used as a preservative to prevent dried fruit from discoloring. It finds application as a sulfonation and sulfomethylation agent in the chemical industry. It is involved in the production of sodium thiosulfate. Other applications include forth flotation ores, oil recovery and manufacture of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500°C (decomposition)
Physical Form Granules
Quantity 10 kg
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,liquid chlorine and liquid ammonia.
Formula Weight 126.04
Percent Purity ≥98%
Grade ACS
Packaging Plastic bottle
Sensitivity Air and moisture sensitive
Specific Gravity 2.633
Density 2.633
Chemical Name or Material Sodium sulfite
Show More Show Less
EINECSNumber 231-821-4
RTECSNumber WE2150000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only