missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, 98+%, ACS reagent, anhydrous

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol

Supplier:  Thermo Scientific Chemicals 424430025

 View more versions of this product

Catalog No. AC424430025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Sodium sulfite
7757-83-7
Fine Crystalline Powder
98% min. (Iodimetry)
Na2SO3
02,387
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
GEHJYWRUCIMESM-UHFFFAOYSA-L
disodium;sulfite
24437
126.04
ACS Reagent
2.63
2.6300g/mL
>500.0°C
White
2.5 kg
Na2O3S
MFCD00003503
15, 8811
Solubility in water: 230 g/mL (20°C). Other solubilities: soluble in glycerol, ppractically insoluble in alcohol
[O-]S(=O)[O-].[Na+].[Na+]
126.04
CHEBI:86477
≥98%
Plastic bottle
>500°C

Chemical Identifiers

7757-83-7
126.04
GEHJYWRUCIMESM-UHFFFAOYSA-L
24437
disodium;sulfite
Na2O3S
MFCD00003503
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
CHEBI:86477
[O-]S(=O)[O-].[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Contact with acids liberates toxic gas.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 231-821-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only