missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, 98.5%, for analysis, anhydrous

Catalog No. AC219270250
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219270250 25 g Glass Bottle
AC219272500 250 g Plastic bottle
AC219270010 1 kg Plastic bottle
AC219270050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC219270250 Supplier Thermo Scientific Chemicals Supplier No. 219270250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500.0°C
Color White
Physical Form Fine Crystals or Crystalline Powder
Quantity 25 g
Packaging Glass Bottle
Specific Gravity 2.63
Assay Percent Range 98% min. (Iodimetry)
Density 2.6300g/mL
Linear Formula Na2SO3
Fieser 02,387
Merck Index 15, 8811
Solubility Information Solubility in water: 230 g/mL (20°C). Other solubilities: soluble in glycerol, ppractically insoluble in alcohol
Formula Weight 126.04
Percent Purity 98.5%
Grade Analytical
Chemical Name or Material Sodium sulfite, Anhydrous, For analysis
Show More Show Less
Hazard Statement GHS H Statement:
Contact with acids liberates toxic gas.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-821-4

RUO – Research Use Only