missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfite, ≥98.0%, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥98.0%
$147.18 - $147.18
Chemical Identifiers
| CAS | 7757-83-7 |
|---|---|
| Molecular Formula | Na2O3S |
| Molecular Weight (g/mol) | 126.037 |
| MDL Number | MFCD00003503 |
| InChI Key | GEHJYWRUCIMESM-UHFFFAOYSA-L |
| Synonym | sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit |
| PubChem CID | 24437 |
| ChEBI | CHEBI:86477 |
| IUPAC Name | disodium;sulfite |
| SMILES | [O-]S(=O)[O-].[Na+].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7757-83-7 | |
| 126.037 | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| 24437 | |
| disodium;sulfite |
| Na2O3S | |
| MFCD00003503 | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| CHEBI:86477 | |
| [O-]S(=O)[O-].[Na+].[Na+] |
Specifications
| 600°C | |
| White | |
| Na2O3S | |
| MFCD00003503 | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| disodium;sulfite | |
| 24437 | |
| 126.04g/mol | |
| ACS Reagent | |
| Powder or Crystals | |
| Sodium Sulfite |
| 7757-83-7 | |
| 500 g | |
| Na2SO3 | |
| NONH for all modes of transport | |
| Soluble in water, 126g/L at 20°C | |
| [O-]S(=O)[O-].[Na+].[Na+] | |
| 126.037 | |
| CHEBI:86477 | |
| ≥98.0% | |
| Poly Bottle | |
| 2.6g/cm3 |
Safety and Handling
EINECSNumber : 231-821-4
RTECSNumber : WE2150000