missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, Decahydrate, Certified, 22.2% (w/v), LabChem™

Catalog No. lc249002
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC249002 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC249002 Supplier LabChem Supplier No. LC249002
Only null left

Chemical Identifiers

CAS 7757-82-6 , 7732-18-5
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Color Colorless
CAS Max % 90.2, 9.8
Physical Form Liquid
Quantity 1 L
Linear Formula Na2SO4
Solubility Information Soluble in water
Formula Weight 142.04
Percent Purity 22.2% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds
Chemical Name or Material Sodium Sulfate, Decahydrate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature