missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, Anhydrous, Low in Nitrogen, Spectrum™ Chemical
$312.82 - $2667.56
Chemical Identifiers
| CAS | 7757-82-6 |
|---|---|
| Molecular Formula | Na2O4S |
| Molecular Weight (g/mol) | 142.04 |
| MDL Number | MFCD00003504 |
| InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
| IUPAC Name | disodium sulfate |
| SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18613864
|
Spectrum Chemical Mfg Cor
S1537500GM |
500 g | Amber Glass Bottle |
Each for $312.82
|
|
||||
|
18613865
|
Spectrum Chemical Mfg Cor
S153725KG |
2.5 kg | Amber Glass Bottle |
Each for $889.21
|
|
||||
|
18613866
|
Spectrum Chemical Mfg Cor
S153712KG |
12 kg | Poly Pail |
Each for $2,667.56
|
|
||||
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
| Na2O4S | |
| MFCD00003504 | |
| disodium sulfate |
Specifications
| 7757-82-6 | |
| 5.2 to 9.2 | |
| Na2O4S | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| Reagent | |
| Amber Glass Bottle |
| 1 | |
| 500 g | |
| MFCD00003504 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| 0.005 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.