missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, Anhydrous, Low in Nitrogen, Spectrum™ Chemical

Catalog No. 18613864
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613864 500 g Amber Glass Bottle
18613865 2.5 kg Amber Glass Bottle
18613866 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613864 Supplier Spectrum Chemical Mfg Cor Supplier No. S1537500GM
Only null left

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

pH 5.2 to 9.2
CAS Max % 1
Quantity 500 g
Grade Reagent
Loss on Ignition 0.005
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.