missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, Anhydrous, Granular, Reagent, ACS, 99%, Spectrum™ Chemical
S1450, 7757-82-6, Na2SO4
$284.33 - $5009.27
Chemical Identifiers
| CAS | 7757-82-6 |
|---|---|
| Molecular Formula | Na2O4S |
| Molecular Weight (g/mol) | 142.04 |
| MDL Number | MFCD00003504 |
| InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
| IUPAC Name | disodium sulfate |
| SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606825
|
Spectrum Chemical Mfg Cor
S1450500GM |
500 g | Amber Glass Bottle |
Each for $284.33
|
|
||||
|
18606827
|
Spectrum Chemical Mfg Cor
S145025KGGL |
2.5 kg | Amber Glass Bottle |
Each for $457.47
|
|
||||
|
18606826
|
Spectrum Chemical Mfg Cor
S1450500GMCS6 |
6 x 500 g | Amber Glass Bottle |
Case of 6 Each for $1,461.37
|
|
||||
|
18606828
|
Spectrum Chemical Mfg Cor
S145025KGGLCS4 |
4 x 2.5 kg | Amber Glass Bottle |
Case of 4 Each for $2,075.80
|
|
||||
|
18606829
|
Spectrum Chemical Mfg Cor
S145012KG |
12 kg | Poly Pail |
Each for $2,306.47
|
|
||||
|
18606830
|
Spectrum Chemical Mfg Cor
S145050KGBL |
50 kg | Fiber Drum |
Each for $5,009.27
|
|
||||
Description
Spectrum™ Chemical Sodium Sulfate, Anhydrous, Granular, Reagent, ACS is the sodium salt of sulfuric acid. It is a white, crystalline solid, and is used on a small scale in laboratories as an inert drying agent. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
| Na2O4S | |
| MFCD00003504 | |
| disodium sulfate |
Specifications
| 7757-82-6 | |
| 5.2 to 9.2 | |
| Na2O4S | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| 99% | |
| 0.005 |
| 1 | |
| 500 g | |
| MFCD00003504 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| ACS | |
| Amber Glass Bottle |