missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, Anhydrous, Granular, Reagent, ACS, 99%, Spectrum™ Chemical
S1450, 7757-82-6, Na2SO4
Supplier: Spectrum Chemical Mfg Cor S1450500GMCS6
Description
Spectrum™ Chemical Sodium Sulfate, Anhydrous, Granular, Reagent, ACS is the sodium salt of sulfuric acid. It is a white, crystalline solid, and is used on a small scale in laboratories as an inert drying agent. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.Specifications
| 7757-82-6 | |
| 5.2 to 9.2 | |
| Na2O4S | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| 99% | |
| Amber Glass Bottle |
| 1 | |
| 6 x 500 g | |
| MFCD00003504 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| 0.005 |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
| Na2O4S | |
| MFCD00003504 | |
| disodium sulfate |