missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, Anhydrous, EMSURE™, MilliporeSigma™
Supplier: Technidata 1066499025
Specifications
| 888°C | |
| 7757-82-6 | |
| Solid | |
| Na2O4S | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| 24436 | |
| 142.04 | |
| ACS | |
| 2.70Â g/3 (20Â °C) |
| White | |
| 5.2 to 8.0 (50g/L, H2O, 20°C) | |
| 25 kg | |
| MFCD00003504 | |
| 200g/L | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| CHEBI:32149 | |
| >99.0% | |
| Fiber Carton |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| 24436 | |
| disodium sulfate |
| Na2O4S | |
| MFCD00003504 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| CHEBI:32149 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
Safety and Handling
EINECSNumber : 231-820-9
RTECSNumber : WE1650000