missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, Anhydrous, EMSURE™, MilliporeSigma™

Catalog No. p-7082924
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Carton
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1066490500 500 g Plastic Bottle
M1066491000 1 kg Plastic Bottle
M1066495000 5 kg Plastic Bottle
M1066499025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1066490500 Supplier Technidata Supplier No. 1.06649.0500
Only null left

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 888°C
Color White
pH 5.2 to 8.0 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 500 g
Solubility Information 200g/L
Formula Weight 142.04
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 2.70 g/3 (20 °C)
Show More Show Less
EINECSNumber 231-820-9
RTECSNumber WE1650000