missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, Anhydrous, EMSURE™, MilliporeSigma™

Supplier:  Technidata 1066491000

 View more versions of this product

Catalog No. M1066491000


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

888°C
7757-82-6
Solid
Na2O4S
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
24436
142.04
ACS
2.70 g/3 (20 °C)
White
5.2 to 8.0 (50g/L, H2O, 20°C)
1 kg
MFCD00003504
200g/L
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
>99.0%
Plastic Bottle

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-820-9

RTECSNumber : WE1650000

SDS
Documents

Documents

Promotions

Promotions