missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous, 10-60 Mesh, MilliporeSigma™

$176.70 - $2687.75

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MSX0760E1
View Documents
Technidata
SX0760E1
500 g Plastic Bottle
Each for $176.70
Only null left
 
MSX0760E20
View Documents
Technidata
SX0760E20
45 kg Fiber Drum w/bag
Each for $2,687.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
dihydrogen oxide, dihydrogen monoxide

Specifications

7732-18-5
500 g
dihydrogen oxide, dihydrogen monoxide
Plastic Bottle
Granules
Na2SO4
142.04
Sodium Sulfate, Anhydrous, 10-60 Mesh
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'