missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, 99+%, for analysis, powder, anhydrous

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

Supplier:  Thermo Scientific Chemicals 218750100

 View more versions of this product

Catalog No. AC218750100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>1700.0°C
884.0°C
7757-82-6
5 to 8 (5% soln. at 20°C)
10 kg
Na2O4S
MFCD00003504
15, 8809
Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99%
0.5% max. (800°C)
Sodium sulfate
99%, p.a.
White
Fine Crystalline Powder
99+%
Na2SO4
01,1103
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
24436
142.04
Analytical
Plastic drum

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
[Na+].[Na+].[O-]S([O-])(=O)=O
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0

SIGNAL WORD: CAUTION!

EINECSNumber : 231-820-9

RTECSNumber : WE1650000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only