missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, 99+%, for analysis, powder, anhydrous

Catalog No. p-222005
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218755000 500 g Plastic bottle
AC218750025 2.5 kg Plastic bottle
AC218750100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218755000 Supplier Thermo Scientific Chemicals Supplier No. 218755000
Only null left

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >1700.0°C
Melting Point 884.0°C
Color White
pH 5 to 8 (5% soln. at 20°C)
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
IUPAC Name disodium;sulfate
Formula Weight 142.04
Percent Purity ≥99%
Grade Analytical
Loss on Ignition 0.5% max. (800°C)
Packaging Plastic bottle
Decomposition Information Decomposition Information: >890°C
Density 2.68
Chemical Name or Material Sodium sulfate, 99%, p.a.
Show More Show Less
Hazard Category SIGNAL WORD: CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
EINECSNumber 231-820-9
RTECSNumber WE1650000
TSCA TSCA

RUO – Research Use Only