missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfamate 98.0+%, TCI America™

Catalog No. S011825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S011825G 25 g
S0118500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S011825G Supplier TCI America Supplier No. S011825G

Chemical Identifiers

CAS 13845-18-6
Molecular Formula H2NNaO3S
Molecular Weight (g/mol) 119.07
MDL Number MFCD00059940
InChI Key QDWYPRSFEZRKDK-UHFFFAOYSA-M
Synonym Aminosulfonic Acid Sodium Salt, Sulfamic Acid Sodium Salt
PubChem CID 23700090
IUPAC Name sodium;sulfamate
SMILES NS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 119.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Sulfamate
EINECSNumber (1)-1132
TSCA Yes