missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Succinate, Hexahydrate, Reagent, 97-102%, Spectrum™ Chemical
S1449, 6106-21-4, C4H4Na2O4·6H2O
Supplier: Spectrum Chemical Mfg Cor S1449500GM
Description
Spectrum™ Chemical Sodium Succinate, Hexahydrate, Reagent , also known as succinic acid, is a diprotic and dicarboxylic acid. It is involved in an energy-yielding process known as the citric acid cycle. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.Specifications
| 6106-21-4 | |
| 500 g | |
| ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
| disodium butanedioate hexahydrate | |
| 97 to 102% | |
| Amber Glass Bottle |
| 1 | |
| C4H16Na2O10 | |
| O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O | |
| 270.14 | |
| Reagent |
Chemical Identifiers
| 6106-21-4 | |
| 270.14 | |
| disodium butanedioate hexahydrate |
| C4H16Na2O10 | |
| ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O |