missing translation for 'onlineSavingsMsg'
Learn More

Sodium Succinate, Hexahydrate, Reagent, 97-102%, Spectrum™ Chemical

Catalog No. p-7158277
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18606812 100 g Amber Glass Bottle
18606813 500 g Amber Glass Bottle
18606814 2.5 kg Poly Pail
18606815 12 kg Fiber Drum
18606816 50 kg Fiber Drum
5 options

Catalog No. 18606812

Supplier: Spectrum Chemical Mfg Cor S1449100GM

Only null left

S1449, 6106-21-4, C4H4Na2O4·6H2O

Spectrum™ Chemical Sodium Succinate, Hexahydrate, Reagent , also known as succinic acid, is a diprotic and dicarboxylic acid. It is involved in an energy-yielding process known as the citric acid cycle. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.14
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
IUPAC Name disodium butanedioate hexahydrate
SMILES O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 97 to 102%
Grade Reagent
Packaging Amber Glass Bottle