missing translation for 'onlineSavingsMsg'
Learn More

Sodium Stearyl Fumarate 98.0+%, TCI America™

Supplier:  TCI America S087925G

 View more versions of this product

Catalog No. S087925G


Only null left

Description

Description

Specifications

Specifications

Sodium Stearyl Fumarate
White-Yellow
MFCD00236640
Fumaric Acid Stearyl Ester Sodium Salt, Sodium Octadecyl Fumarate, Fumaric Acid Octadecyl Ester Sodium Salt
CCCCCCCCCCCCCCCCCCOC(=O)C=CC(=O)[O-].[Na+]
390.54
390.54
Crystalline Powder
4070-80-8
C22H39NaO4
25 g
STFSJTPVIIDAQX-UHFFFAOYSA-M
sodium;4-octadecoxy-4-oxobut-2-enoate
53400819
≥98.0% (T)

Chemical Identifiers

4070-80-8
390.54
STFSJTPVIIDAQX-UHFFFAOYSA-M
53400819
CCCCCCCCCCCCCCCCCCOC(=O)C=CC(=O)[O-].[Na+]
C22H39NaO4
MFCD00236640
Fumaric Acid Stearyl Ester Sodium Salt, Sodium Octadecyl Fumarate, Fumaric Acid Octadecyl Ester Sodium Salt
sodium;4-octadecoxy-4-oxobut-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1087

TSCA : No

SDS
Documents

Documents

Promotions

Promotions