missing translation for 'onlineSavingsMsg'
Learn More

Sodium Stearyl Fumarate 98.0+%, TCI America™

Catalog No. p-7135977
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S087925G 25 g
S0879100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S087925G Supplier TCI America Supplier No. S087925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4070-80-8
Molecular Formula C22H39NaO4
Molecular Weight (g/mol) 390.54
MDL Number MFCD00236640
InChI Key STFSJTPVIIDAQX-UHFFFAOYSA-M
Synonym Fumaric Acid Stearyl Ester Sodium Salt, Sodium Octadecyl Fumarate, Fumaric Acid Octadecyl Ester Sodium Salt
PubChem CID 53400819
IUPAC Name sodium;4-octadecoxy-4-oxobut-2-enoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)C=CC(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 390.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Stearyl Fumarate
EINECSNumber (2)-1087
TSCA No