missing translation for 'onlineSavingsMsg'
Learn More

Sodium Stearate, Powder, NF, 90%, Spectrum™ Chemical

Catalog No. 18606798
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606798 2.5 kg Poly Pail
18606799 12 kg Fiber Drum
18606800 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606798 Supplier Spectrum Chemical Mfg Cor Supplier No. S14472.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

S1447, 822-16-2, C18H35NaO2

Spectrum™ Chemical Sodium Stearate, Powder, NF is used in the pharmaceutical industry as a surfactant to aid the solubility of hydrophobic compounds. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 822-16-2
Molecular Formula C18H35NaO2
Molecular Weight (g/mol) 306.47
InChI Key RYYKJJJTJZKILX-UHFFFAOYSA-M
IUPAC Name sodium octadecanoate
SMILES [Na+].CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 90%
Grade NF
Loss on Drying 0.05
Packaging Poly Pail