Learn More
Sodium stearate
CAS: 822-16-2 | C18H35NaO2 | 306.466 g/mol
$93.78 - $563.74
Chemical Identifiers
CAS | 822-16-2 |
---|---|
Molecular Formula | C18H35NaO2 |
Molecular Weight (g/mol) | 306.466 |
MDL Number | MFCD00036404 |
InChI Key | RYYKJJJTJZKILX-UHFFFAOYSA-M |
Synonym | sodium stearate, sodium octadecanoate, octadecanoic acid, sodium salt, stearates, stearic acid, sodium salt, prodhygine, flexichem b, stearic acid sodium salt, bonderlube 235, unii-qu7e2xa9tgShow More |
PubChem CID | 2724691 |
IUPAC Name | sodium;octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA3964830
|
Thermo Scientific Chemicals
3964830 |
250 g |
Each for $93.78
|
|
||||||
AA39648A1
|
Thermo Scientific Chemicals
039648A1 |
1 kg |
Each for $158.19
|
|
||||||
AA39648A7
|
Thermo Scientific Chemicals
39648A7 |
5 kg |
Each for $563.74
|
|
||||||
Description
Used in adhesives and sealants, laundry and dishwashing products, plastic and rubber products. Used as surface active agents. It is the gelling agent for deodrant sticks. Used as waterproofing additives and ointments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
822-16-2 | |
306.466 | |
RYYKJJJTJZKILX-UHFFFAOYSA-M | |
2724691 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].[Na+] |
C18H35NaO2 | |
MFCD00036404 | |
sodium stearate, sodium octadecanoate, octadecanoic acid, sodium salt, stearates, stearic acid, sodium salt, prodhygine, flexichem b, stearic acid sodium salt, bonderlube 235, unii-qu7e2xa9tgShow More | |
sodium;octadecanoate |
Specifications
822-16-2 | |
C18H35NaO2 | |
250 g | |
sodium stearate, sodium octadecanoate, octadecanoic acid, sodium salt, stearates, stearic acid, sodium salt, prodhygine, flexichem b, stearic acid sodium salt, bonderlube 235, unii-qu7e2xa9tg | |
RYYKJJJTJZKILX-UHFFFAOYSA-M | |
sodium;octadecanoate | |
2724691 | |
Powder |
Weak | |
MFCD00036404 | |
14,8678 | |
Slowly soluble in cold water. | |
CCCCCCCCCCCCCCCCCC(=O)[O-].[Na+] | |
306.466 | |
306.46 | |
Sodium stearate |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
EINECSNumber : 212-490-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only