missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Stannate, Granular, Reagent, Spectrum™ Chemical
S1445, 12027-70-2, Na2SnO3·3H2O
$376.10 - $27782.93
Chemical Identifiers
| CAS | 12027-70-2 |
|---|---|
| Molecular Formula | H6Na2O6Sn |
| Molecular Weight (g/mol) | 266.73 |
| InChI Key | PMPBLIIMRRPPEO-UHFFFAOYSA-H |
| IUPAC Name | tin(4+) disodium hexahydroxide |
| SMILES | [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606788
|
Spectrum Chemical Mfg Cor
S1445125GM |
125 g | Amber Glass Bottle |
Each for $376.10
|
|
||||
|
18606789
|
Spectrum Chemical Mfg Cor
S1445500GM |
500 g | Amber Glass Bottle |
Each for $918.83
|
|
||||
|
18606790
|
Spectrum Chemical Mfg Cor
S144525KG |
2.5 kg | Amber Glass Bottle |
Each for $3,135.07
|
|
||||
|
18606791
|
Spectrum Chemical Mfg Cor
S144512KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18606792
|
Spectrum Chemical Mfg Cor
S144545KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sodium Stannate, Granular, Reagent is a colorless salt that occurs when tin is dissolved in sodium hydroxide. It is an inorganic compound that is used as a hydrogen peroxide stabilizer. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Chemical Identifiers
| 12027-70-2 | |
| 266.73 | |
| tin(4+) disodium hexahydroxide |
| H6Na2O6Sn | |
| PMPBLIIMRRPPEO-UHFFFAOYSA-H | |
| [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4] |
Specifications
| 12027-70-2 | |
| 125 g | |
| PMPBLIIMRRPPEO-UHFFFAOYSA-H | |
| tin(4+) disodium hexahydroxide | |
| Reagent |
| 1 | |
| H6Na2O6Sn | |
| [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4] | |
| 266.73 | |
| Amber Glass Bottle |