missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Stannate, Granular, Reagent, Spectrum™ Chemical
S1445, 12027-70-2, Na2SnO3·3H2O
Supplier: Spectrum Chemical Mfg Cor S144525KG
Description
Spectrum™ Chemical Sodium Stannate, Granular, Reagent is a colorless salt that occurs when tin is dissolved in sodium hydroxide. It is an inorganic compound that is used as a hydrogen peroxide stabilizer. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Specifications
| 12027-70-2 | |
| 2.5 kg | |
| PMPBLIIMRRPPEO-UHFFFAOYSA-H | |
| tin(4+) disodium hexahydroxide | |
| Reagent |
| 1 | |
| H6Na2O6Sn | |
| [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4] | |
| 266.73 | |
| Amber Glass Bottle |
Chemical Identifiers
| 12027-70-2 | |
| 266.73 | |
| tin(4+) disodium hexahydroxide |
| H6Na2O6Sn | |
| PMPBLIIMRRPPEO-UHFFFAOYSA-H | |
| [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4] |