missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard Solution, METTLER TOLEDO™

Catalog No. 01912574
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01912574 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01912574 Supplier METTLER TOLEDO Supplier No. 30097048

No weighing or dilution is required

  • Sodium Standard
  • 2000 ppm (0.5 M)
  • No weighing or dilution is required

Specifications

Chemical Name or Material Sodium Standard 2000 ppm
Packaging Plastic Bottle
CAS 7647-14-5, 7732-18-5
Molecular Formula NaCl, H2O
Concentration 2000 ppm (0.5 M)
Color Colorless
pH 7
Melting Point 0°C
Boiling Point 100°C
Synonym None
Type Analytical Standard
Molecular Weight (g/mol) 58.44
Quantity 1 L
Show More Show Less