missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLNA22X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Na | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Sodium Standard | |
| 7440-23-5,7732-18-5,7697-37-2 | |
| Na | |
| AA, ICP | |
| Certified Reference Material | |
| [Na] | |
| sodium | |
| 500 mL | |
| CHEBI:26708 |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00085307 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| KEAYESYHFKHZAL-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 22.99 | |
| 5360545 | |
| Assurance |
Product intended for laboratory use only