missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicofluoride, 95%, Spectrum™ Chemical

$218.68 - $7214.75

Chemical Identifiers

CAS 16893-85-9
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613857
View Documents
Spectrum Chemical Mfg Cor
S144025GM
25 g Poly Bottle
Each for $218.68
Only null left
 
18613858
View Documents
Spectrum Chemical Mfg Cor
S1440100GM
100 g Poly Bottle
Each for $466.29
Only null left
 
18613860
View Documents
Spectrum Chemical Mfg Cor
S1440500GM
500 g Poly Bottle
Each for $1,261.54
Only null left
 
18613861
View Documents
Spectrum Chemical Mfg Cor
S1440100LBBL
100 lb. Fiber Drum
Each for $7,214.75
Only null left
 
18613862
View Documents
Spectrum Chemical Mfg Cor
S144025KG
2.5 kg Poly Bottle
Each for $2,132.36
Only null left
 
18613859
View Documents
Spectrum Chemical Mfg Cor
S144012KG
12 kg Poly Pail
Each for $2,737.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16893-85-9

Specifications

16893-85-9
25 g
Reagent
Poly Bottle
1
95%
0.005
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.