missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicofluoride, 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1440100LBBL

 View more versions of this product

Catalog No. 18613861


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

16893-85-9
100 lb.
Reagent
Fiber Drum
1
95%
0.005

Chemical Identifiers

16893-85-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.