missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate, Technical Grade, 40 to 42°Baumé, Ricca Chemical

Catalog No. R75550001A
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R75550001A 1 L Natural Poly Bottle
75551 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R75550001A Supplier Ricca Chemical Company Supplier No. R75550001A
Only null left

Chemical Identifiers

CAS 7732-18-5 , 1344-09-8
Molecular Formula H10Na2O8Si
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium oxosilanebis(olate) pentahydrate
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Color Colorless or White
Physical Form Liquid
Quantity 1 L
Solubility Information Solubility in Water: Infinite
Molecular Weight (g/mol) 212.14
Grade Technical
Odor Odorless
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Silicate, 40 to 42°Be