missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sesquicarbonate, Dihydrate, Spectrum™ Chemical
S1357, 533-96-0, Na2CO3·NaHCO3·2H2O
Supplier: Spectrum Chemical Mfg Cor S135725KG
Description
Spectrum™ Chemical Sodium Sesquicarbonate, Dihydrate , with a systematic name trisodium hydrogen dicarbonate, is used as an alkalinity source for water treatment. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 533-96-0 | |
| 2.5 kg | |
| WCTAGTRAWPDFQO-UHFFFAOYSA-K | |
| trisodium hydrogen carbonate carbonate | |
| Reagent |
| 1 | |
| C2HNa3O6 | |
| [Na+].[Na+].[Na+].OC([O-])=O.[O-]C([O-])=O | |
| 189.99 | |
| Poly Bottle |
Chemical Identifiers
| 533-96-0 | |
| 189.99 | |
| trisodium hydrogen carbonate carbonate |
| C2HNa3O6 | |
| WCTAGTRAWPDFQO-UHFFFAOYSA-K | |
| [Na+].[Na+].[Na+].OC([O-])=O.[O-]C([O-])=O |