accessibility menu, dialog, popup

UserName

CAS RN 533-96-0

NaNaNaOHOOOOO

CAS RN 533-96-0

IUPAC Name: trisodium hydrogen carbonate carbonate
Synonyms: sodium sesquicarbonate
Molecular Weight (g/mol): 189.99
Molecular Formula: C2HNa3O6
InChi Key: WCTAGTRAWPDFQO-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].OC([O-])=O.[O-]C([O-])=O
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)