missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sesquicarbonate, Dihydrate, Spectrum™ Chemical

Catalog No. 18606784
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606784 500 g Poly Bottle
18606785 2.5 kg Poly Bottle
18606786 12 kg Poly Pail
18606787 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606784 Supplier Spectrum Chemical Mfg Cor Supplier No. S1357500GM
Only null left

S1357, 533-96-0, Na2CO3·NaHCO3·2H2O

Spectrum™ Chemical Sodium Sesquicarbonate, Dihydrate , with a systematic name trisodium hydrogen dicarbonate, is used as an alkalinity source for water treatment. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 533-96-0
Molecular Formula C2HNa3O6
Molecular Weight (g/mol) 189.99
InChI Key WCTAGTRAWPDFQO-UHFFFAOYSA-K
IUPAC Name trisodium hydrogen carbonate carbonate
SMILES [Na+].[Na+].[Na+].OC([O-])=O.[O-]C([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Grade Reagent
Packaging Poly Bottle