missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Selenate, Anhydrous, Crystal, 97%, Spectrum™ Chemical
S1428, 13410-01-0, Na2SeO4
Supplier: Spectrum Chemical Mfg Cor S1428100GMGL
Description
Spectrum™ Chemical Sodium Selenate, Anhydrous, Crystal has several uses, including the manufacturing of glass, specifically red-hued glass, but can also produce clear glass when mixed with ferrous oxides. It's a white or gray solid substance that is highly toxic. Regardless of the toxicity, sodium selenate is used in vitamin supplements as a trace mineral additive and is being investigated as a potential anti-Alzheimers drug candidate.Specifications
| 13410-01-0 | |
| 100 g | |
| MFCD00003490 | |
| [Na+].[Na+].[O-][Se]([O-])(=O)=O | |
| 188.95 | |
| Reagent |
| 1 | |
| Na2O4Se | |
| MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
| disodium selenate | |
| 97% | |
| Amber Glass Bottle |
Chemical Identifiers
| 13410-01-0 | |
| 188.95 | |
| MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-][Se]([O-])(=O)=O |
| Na2O4Se | |
| MFCD00003490 | |
| disodium selenate |