missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenate, Anhydrous, Crystal, 97%, Spectrum™ Chemical

Catalog No. 18606779
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606779 25 g Amber Glass Bottle
18606780 100 g Amber Glass Bottle
18606781 500 g Amber Glass Bottle
18606782 5 kg Poly Pail
18606783 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606779 Supplier Spectrum Chemical Mfg Cor Supplier No. S142825GM

S1428, 13410-01-0, Na2SeO4

Spectrum™ Chemical Sodium Selenate, Anhydrous, Crystal has several uses, including the manufacturing of glass, specifically red-hued glass, but can also produce clear glass when mixed with ferrous oxides. It's a white or gray solid substance that is highly toxic. Regardless of the toxicity, sodium selenate is used in vitamin supplements as a trace mineral additive and is being investigated as a potential anti-Alzheimers drug candidate.

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
IUPAC Name disodium selenate
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 97%
Grade Reagent
Packaging Amber Glass Bottle