missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, Reagent Grade, Reagents

Catalog No. p-200021166
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337685 125 g
01337684 2.5 kg
01337683 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337685 Supplier Labchem Inc Supplier No. C1324700125D2
Only null left

Sodium Salicylate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym Sodium 2-hydroxybenzoate
IUPAC Name sodium 2-hydroxybenzoate
SMILES [Na+].OC1=CC=CC=C1C([O-])=O

Specifications

Boiling Point 336°C
Color White or Slightly Yellow
Grade Reagent
Quantity 125 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 0.32
Chemical Name or Material Sodium Salicylate, Reagent Grade