missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, Reagent Grade, Reagents

Catalog No. 01337685
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337685 125 g
01337684 2.5 kg
01337683 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337685 Supplier Labchem Inc Supplier No. C1324700125D2

Sodium Salicylate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym Sodium 2-hydroxybenzoate
IUPAC Name sodium 2-hydroxybenzoate
SMILES [Na+].OC1=CC=CC=C1C([O-])=O

Specifications

Boiling Point 336°C
Color White or Slightly Yellow
Grade Reagent
Quantity 125 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 0.32
Chemical Name or Material Sodium Salicylate, Reagent Grade