missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. p-7082923
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1066010250 250 g Plastic Bottle
M1066011000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1066010250 Supplier Technidata Supplier No. 1.06601.0250
Only null left

Recommended Storage

+5°C to +30°C.

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.104
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
PubChem CID 16760658
ChEBI CHEBI:9180
IUPAC Name sodium;2-hydroxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]

Specifications

Melting Point 250°C (decomposition)
Color White
pH 8.7 to 9.3 (10g/L, H2O, 20°C)
Physical Form Crystals
Quantity 250 g
Solubility Information 77g/L
Formula Weight 141.96
Percent Purity >99.0%
Packaging Plastic Bottle
Density 0.32g/cm3 (20°C)
EINECSNumber 200-198-0
RTECSNumber VO5075000
Recommended Storage +5°C to +30°C.