missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, Crystal, 99%, Spectrum™ Chemical

Catalog No. 18613841
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613841 125 g Amber Glass Bottle
18613842 500 g Amber Glass Bottle
18613843 2.5 kg Poly Pail
18613844 12 kg Poly Pail
18613845 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613841 Supplier Spectrum Chemical Mfg Cor Supplier No. S1425125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 54-21-7

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.