missing translation for 'onlineSavingsMsg'
Learn More

Sodium Saccharin, Dihydrate, Granular, FCC, 98-101%, Spectrum™ Chemical

Catalog No. p-7158267
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606771 100 g Poly Bottle
18606768 500 g Poly Bottle
18606769 2.5 kg Poly Pail
18606770 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606771 Supplier Spectrum Chemical Mfg Cor Supplier No. S1421100GM
Only null left

S1421, 6155-57-3, C7H4NNaO3S·2H2O

Spectrum™ Chemical Sodium Saccharin, Dihydrate, Granular, FCC is a white crystalline solid whose most common use is as an artificial sweetener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6155-57-3
Molecular Formula C7H8NNaO5S
Molecular Weight (g/mol) 241.19
InChI Key NAUHOSQQKJTUAX-UHFFFAOYSA-M
IUPAC Name sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate dihydrate
SMILES O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Percent Purity 98 to 101%
Grade FCC